Assay Details
| Gene Symbol: abi2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076586.2 | NP_001070054.2 | 4 - 5 | 557 | 96 | |
| XM_005167480.4 | XP_005167537.1 | 4 - 5 | 584 | 96 | ||
| XM_005167483.4 | XP_005167540.1 | 4 - 5 | 584 | 96 | ||
| XM_009304512.3 | XP_009302787.1 | 5 - 6 | 632 | 96 | ||
| XM_009304513.3 | XP_009302788.1 | 4 - 5 | 584 | 96 | ||
| XM_009304514.3 | XP_009302789.1 | 5 - 6 | 631 | 96 | ||
| XM_009304515.3 | XP_009302790.1 | 5 - 6 | 631 | 96 | ||
| XM_009304516.3 | XP_009302791.1 | 5 - 6 | 630 | 96 | ||
| XM_017357719.2 | XP_017213208.1 | 5 - 6 | 632 | 96 | ||
| XM_017357720.2 | XP_017213209.1 | 5 - 6 | 632 | 96 | ||
| XM_021478762.1 | XP_021334437.1 | 5 - 6 | 631 | 96 | ||
| XM_021478763.1 | XP_021334438.1 | 4 - 5 | 584 | 96 | ||
| XM_021478764.1 | XP_021334439.1 | 4 - 5 | 587 | 96 | ||
| GenBank mRNA | BC122368.1 | - | 4 - 5 | 593 | 96 | |
| BC165556.1 | - | 4 - 5 | 591 | 96 | ||
More Information
Set Membership: |
|

abi2a
Genomic Map