Assay Details
| Gene Symbol: agpat5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076745.2 | NP_001070213.2 | 7 - 8 | 1221 | 73 | |
| GenBank mRNA | BC124532.1 | - | 7 - 8 | 1016 | 73 | |
| BC164409.1 | - | 7 - 8 | 907 | 73 | ||
More Information
Set Membership: |
|

agpat5
Genomic Map