Assay Details
| Gene Symbol: ecsit | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242938.1 | NP_001229867.1 | 6 - 7 | 1133 | 74 | |
| XM_009302997.3 | XP_009301272.1 | 6 - 7 | 1157 | 74 | ||
| GenBank mRNA | BC124208.1 | - | 6 - 7 | 1093 | 74 | |
| BC164678.1 | - | 5 - 6 | 1019 | 74 | ||
| DY551107.1 | - | 4 - 5 | 633 | 74 | ||
| EH600372.1 | - | 2 - 3 | 635 | 74 | ||
More Information
Set Membership: |
|

ecsit
Genomic Map