Assay Details
| Gene Symbol: mtfr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077285.1 | NP_001070753.1 | 7 - 8 | 1120 | 80 | |
| GenBank mRNA | BC124770.1 | - | 7 - 8 | 1120 | 80 | |
More Information
Set Membership: |
|

mtfr2
Genomic Map