Assay Details
| Gene Symbol: cttnbp2nlb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077371.1 | NP_001070839.1 | 3 - 4 | 286 | 81 | |
| XM_005167035.4 | XP_005167092.1 | 3 - 4 | 297 | 81 | ||
| GenBank mRNA | BC124670.1 | - | 3 - 4 | 286 | 81 | |
More Information
Set Membership: |
|

cttnbp2nlb
Genomic Map