Assay Details
| Gene Symbol: otud7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077744.1 | NP_001071212.1 | 11 - 12 | 1500 | 72 | |
| XM_005158389.4 | XP_005158446.1 | 11 - 12 | 1613 | 72 | ||
| GenBank mRNA | BC125816.1 | - | 11 - 12 | 1500 | 72 | |
More Information
Set Membership: |
|

otud7b
Genomic Map