Assay Details
| Gene Symbol: tmem41ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213083.1 | NP_998248.1 | 4 - 5 | 649 | 83 | |
| XM_021469451.1 | XP_021325126.1 | 4 - 5 | 540 | 83 | ||
| GenBank mRNA | BC068328.1 | - | 4 - 5 | 649 | 83 | |
More Information
Set Membership: |
|

tmem41ab
Genomic Map