Assay Details
| Gene Symbol: nsd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082551.1 | NP_001076020.1 | 21 - 22 | 4041 | 76 | |
| GenBank mRNA | BC093421.1 | - | 3 - 4 | 366 | 76 | |
| BC115211.1 | - | 6 - 7 | 862 | 76 | ||
| BC171696.1 | - | 21 - 22 | 4035 | 76 | ||
| DQ358102.1 | - | 20 - 21 | 3504 | 76 | ||
More Information
Set Membership: |
|

nsd2
Genomic Map