Assay Details
| Gene Symbol: pik3r6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083008.1 | NP_001076477.1 | 3 - 4 | 380 | 140 | |
| XM_009302370.3 | XP_009300645.1 | 3 - 4 | 519 | 140 | ||
| GenBank mRNA | BC133142.1 | - | 3 - 4 | 380 | 140 | |
More Information
Set Membership: |
|

pik3r6a
Genomic Map