Assay Details
| Gene Symbol: usp12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083556.1 | NP_001077025.1 | 8 - 9 | 1034 | 114 | |
| GenBank mRNA | BC115286.1 | - | 8 - 9 | 1034 | 114 | |
| EG583233.1 | - | 3 - 4 | 307 | 114 | ||
More Information
Set Membership: |
|

usp12a
Genomic Map