Assay Details
| Gene Symbol: map2k4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089421.1 | NP_001082890.1 | 8 - 9 | 740 | 98 | |
| XM_005172951.4 | XP_005173008.1 | 7 - 8 | 1091 | 98 | ||
| XM_021480155.1 | XP_021335830.1 | 8 - 9 | 1128 | 98 | ||
| XM_021480156.1 | XP_021335831.1 | 7 - 8 | 1096 | 98 | ||
| GenBank mRNA | AB438979.1 | - | 8 - 9 | 740 | 98 | |
| BC162777.1 | - | 8 - 9 | 816 | 98 | ||
| EF489398.1 | - | 8 - 9 | 740 | 98 | ||
More Information
Set Membership: |
|

map2k4b
Genomic Map