Assay Details
| Gene Symbol: syt1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089461.1 | NP_001082930.1 | 4 - 5 | 651 | 77 | |
| XM_021467559.1 | XP_021323234.1 | 5 - 6 | 991 | 77 | ||
| XM_021467560.1 | XP_021323235.1 | 4 - 5 | 1052 | 77 | ||
| Gene Symbol: LOC103909492 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009295707.2 | XP_009293982.1 | 3 - 4 | 390 | 77 | |
More Information
Set Membership: |
|

syt1b
Genomic Map