Assay Details
| Gene Symbol: syt1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089461.1 | NP_001082930.1 | 5 - 6 | 819 | 64 | |
| XM_021467559.1 | XP_021323234.1 | 6 - 7 | 1159 | 64 | ||
| XM_021467560.1 | XP_021323235.1 | 5 - 6 | 1220 | 64 | ||
More Information
Set Membership: |
|

syt1b
Genomic Map