Assay Details
| Gene Symbol: syt1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089461.1 | NP_001082930.1 | 7 - 8 | 1068 | 139 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

syt1b| Gene Symbol: syt1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089461.1 | NP_001082930.1 | 7 - 8 | 1068 | 139 | |
Set Membership: |
|