Assay Details
| Gene Symbol: yme1l1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331068.1 | NP_001317997.1 | 5 - 6 | 643 | 71 | |
| GenBank mRNA | BC139529.1 | - | 5 - 6 | 643 | 71 | |
More Information
Set Membership: |
|

yme1l1b
Genomic Map