Assay Details
| Gene Symbol: yme1l1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331068.1 | NP_001317997.1 | 6 - 7 | 76 | ||
| GenBank mRNA | BC139529.1 | - | 6 - 7 | 786 | 76 | |
More Information
Set Membership: |
|

yme1l1b
Genomic Map