Assay Details
| Gene Symbol: yme1l1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331068.1 | NP_001317997.1 | 8 - 9 | 1042 | 61 | |
| GenBank mRNA | BC139529.1 | - | 8 - 9 | 1042 | 61 | |
More Information
Set Membership: |
|

yme1l1b
Genomic Map