Assay Details
| Gene Symbol: oxsr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098747.1 | NP_001092217.1 | 13 - 14 | 1367 | 57 | |
| GenBank mRNA | BC124256.1 | - | 13 - 14 | 1414 | 57 | |
| BC142933.1 | - | 13 - 14 | 1367 | 57 | ||
More Information
Set Membership: |
|

oxsr1a
Genomic Map