Assay Details
| Gene Symbol: oxsr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098747.1 | NP_001092217.1 | 17 - 18 | 1626 | 134 | |
| GenBank mRNA | BC124256.1 | - | 17 - 18 | 1673 | 134 | |
| BC142933.1 | - | 17 - 18 | 1626 | 134 | ||
More Information
Set Membership: |
|

oxsr1a
Genomic Map