Assay Details
| Gene Symbol: loxl2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099244.1 | NP_001092714.1 | 5 - 6 | 1056 | 74 | |
| XM_009305250.3 | XP_009303525.1 | 6 - 7 | 1242 | 74 | ||
| GenBank mRNA | EF030481.1 | - | 5 - 6 | 1056 | 74 | |
More Information
Set Membership: |
|

loxl2a
Genomic Map