Assay Details
| Gene Symbol: zswim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100143.1 | NP_001093613.1 | 1 - 2 | 301 | 64 | |
| GenBank mRNA | BC146750.1 | - | 1 - 2 | 301 | 64 | |
More Information
Set Membership: |
|

zswim2
Genomic Map