Assay Details
| Gene Symbol: npr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102673.1 | NP_001096143.1 | 7 - 8 | 1716 | 82 | |
| XM_005165356.4 | XP_005165413.1 | 7 - 8 | 1762 | 82 | ||
| GenBank mRNA | BC133098.1 | - | 7 - 8 | 1716 | 82 | |
More Information
Set Membership: |
|

npr3
Genomic Map