Assay Details
| Gene Symbol: kiss1ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105679.2 | NP_001099149.2 | 3 - 4 | 485 | 78 | |
| GenBank mRNA | AB623211.1 | - | 3 - 4 | 485 | 78 | |
| EF672263.1 | - | 2 - 3 | 263 | 78 | ||
| EU047917.1 | - | 3 - 4 | 485 | 78 | ||
| HQ222888.1 | - | 3 - 4 | 485 | 78 | ||
More Information
Set Membership: |
|

kiss1ra
Genomic Map