Assay Details
| Gene Symbol: dync2i2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105690.1 | NP_001099160.1 | 3 - 4 | 776 | 72 | |
| XM_021468923.1 | XP_021324598.1 | 3 - 4 | 795 | 72 | ||
| GenBank mRNA | BC095866.1 | - | 3 - 4 | 774 | 72 | |
| BC133909.1 | - | 3 - 4 | 381 | 72 | ||
| BC152526.1 | - | 3 - 4 | 776 | 72 | ||
More Information
Set Membership: |
|

dync2i2
Genomic Map