Assay Details
| Gene Symbol: ddx3xb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130941.2 | NP_571016.2 | 16 - 17 | 2105 | 78 | |
| XM_005168788.4 | XP_005168845.1 | 17 - 18 | 2257 | 78 | ||
| XM_005168789.4 | XP_005168846.1 | 17 - 18 | 2257 | 78 | ||
| XM_005168790.4 | XP_005168847.1 | 17 - 18 | 2209 | 78 | ||
| XM_005168791.4 | XP_005168848.1 | 16 - 17 | 2206 | 78 | ||
| XM_005168792.4 | XP_005168849.1 | 16 - 17 | 2206 | 78 | ||
| GenBank mRNA | BC059794.1 | - | 16 - 17 | 2105 | 78 | |
| Y12819.1 | - | 2110 | 78 | |||
More Information
Set Membership: |
|

ddx3xb
Genomic Map