Assay Details
| Gene Symbol: ddx3xb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130941.2 | NP_571016.2 | 17 - 18 | 2232 | 66 | |
| XM_005168788.4 | XP_005168845.1 | 18 - 19 | 2384 | 66 | ||
| XM_005168790.4 | XP_005168847.1 | 18 - 19 | 2336 | 66 | ||
| XM_005168791.4 | XP_005168848.1 | 17 - 18 | 2333 | 66 | ||
| GenBank mRNA | BC059794.1 | - | 17 - 18 | 2232 | 66 | |
| Y12819.1 | - | 2237 | 66 | |||
More Information
Set Membership: |
|

ddx3xb
Genomic Map