Assay Details
| Gene Symbol: egr2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130997.2 | NP_571072.2 | 1 - 2 | 314 | 96 | |
| GenBank mRNA | BC081622.1 | - | 1 - 2 | 314 | 96 | |
| X70322.1 | - | 200 | 96 | |||
More Information
Set Membership: |
|

egr2b
Genomic Map