Assay Details
| Gene Symbol: en2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131044.2 | NP_571119.2 | 1 - 2 | 794 | 71 | |
| GenBank mRNA | BC162325.1 | - | 1 - 2 | 505 | 71 | |
| BC162618.1 | - | 1 - 2 | 505 | 71 | ||
| X68151.1 | - | 795 | 71 | |||
| X68446.1 | - | 519 | 71 | |||
More Information
Set Membership: |
|

en2a
Genomic Map