Assay Details
Gene Symbol: tbx16l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131052.2 | NP_571127.2 | 2 - 3 | 222 | 116 | |
XM_017356035.2 | XP_017211524.1 | 2 - 3 | 702 | 116 | ||
XM_017356037.2 | XP_017211526.1 | 2 - 3 | 707 | 116 | ||
GenBank mRNA | BC099989.1 | - | 2 - 3 | 195 | 116 | |
U80951.1 | - | 2 - 3 | 231 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
