Assay Details
| Gene Symbol: tbx16l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131052.2 | NP_571127.2 | 5 - 6 | 645 | 63 | |
| XM_017356035.2 | XP_017211524.1 | 5 - 6 | 1125 | 63 | ||
| XM_017356037.2 | XP_017211526.1 | 5 - 6 | 1130 | 63 | ||
| GenBank mRNA | BC099989.1 | - | 5 - 6 | 618 | 63 | |
| U80951.1 | - | 5 - 6 | 654 | 63 | ||
More Information
Set Membership: |
|

tbx16l
Genomic Map