Assay Details
| Gene Symbol: tcp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131230.1 | NP_571305.1 | 7 - 8 | 809 | 67 | |
| GenBank mRNA | AF143493.1 | - | 7 - 8 | 809 | 67 | |
| AF164028.1 | - | 6 - 7 | 747 | 67 | ||
| AF164029.1 | - | 4 - 5 | 527 | 67 | ||
| AF164030.1 | - | 4 - 5 | 527 | 67 | ||
| BC044397.1 | - | 7 - 8 | 867 | 67 | ||
| BC066538.1 | - | 7 - 8 | 809 | 67 | ||
| BC164034.1 | - | 7 - 8 | 845 | 67 | ||
More Information
Set Membership: |
|

tcp1
Genomic Map