Assay Details
| Gene Symbol: tcf7l1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131269.2 | NP_571344.1 | 11 - 12 | 1768 | 78 | |
| GenBank mRNA | AF097410.1 | - | 11 - 12 | 1346 | 78 | |
| BC053135.1 | - | 11 - 12 | 1768 | 78 | ||
More Information
Set Membership: |
|

tcf7l1a
Genomic Map