Assay Details
| Gene Symbol: gfap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131373.2 | NP_571448.2 | 4 - 5 | 840 | 67 | |
| XM_005163983.4 | XP_005164040.1 | 5 - 6 | 1084 | 67 | ||
| GenBank mRNA | AF506734.1 | - | 4 - 5 | 827 | 67 | |
| AY151283.1 | - | 4 - 5 | 819 | 67 | ||
| AY151284.1 | - | 4 - 5 | 819 | 67 | ||
| AY397679.1 | - | 4 - 5 | 1135 | 67 | ||
| BC068410.1 | - | 4 - 5 | 817 | 67 | ||
| BC091942.1 | - | 4 - 5 | 840 | 67 | ||
| BC128803.1 | - | 4 - 5 | 817 | 67 | ||
| BC155310.1 | - | 4 - 5 | 815 | 67 | ||
| BC164211.1 | - | 4 - 5 | 853 | 67 | ||
| FJ915067.1 | - | 1 - 2 | 42 | 67 | ||
More Information
Set Membership: |
|

gfap
Genomic Map