Assay Details
| Gene Symbol: igfbp2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131458.2 | NP_571533.1 | 2 - 3 | 643 | 133 | |
| GenBank mRNA | AF198033.1 | - | 2 - 3 | 645 | 133 | |
| BC056331.1 | - | 2 - 3 | 652 | 133 | ||
| BC065627.1 | - | 2 - 3 | 629 | 133 | ||
| BC153606.1 | - | 2 - 3 | 564 | 133 | ||
More Information
Set Membership: |
|

igfbp2a
Genomic Map