Assay Details
| Gene Symbol: hoxa4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131535.1 | NP_571610.1 | 1 - 2 | 468 | 86 | |
| GenBank mRNA | BC122457.1 | - | 1 - 2 | 468 | 86 | |
More Information
Set Membership: |
|

hoxa4a
Genomic Map