Assay Details
| Gene Symbol: badb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270595.1 | NP_001257524.1 | 2 - 3 | 228 | 80 | |
| NM_131579.2 | NP_571654.1 | 2 - 3 | 228 | 80 | ||
| XM_005172305.4 | XP_005172362.1 | 2 - 3 | 235 | 80 | ||
| GenBank mRNA | AF231017.2 | - | 2 - 3 | 192 | 80 | |
| BC152237.1 | - | 2 - 3 | 187 | 80 | ||
| BC164947.1 | - | 1 - 2 | 173 | 80 | ||
| EH498043.1 | - | 2 - 3 | 256 | 80 | ||
| EH592357.1 | - | 2 - 3 | 256 | 80 | ||
More Information
Set Membership: |
|

badb
Genomic Map