Assay Details
| Gene Symbol: mcl1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131599.1 | NP_571674.1 | 1 - 2 | 415 | 91 | |
| GenBank mRNA | AF231016.1 | - | 1 - 2 | 407 | 91 | |
| AF302805.1 | - | 1 - 2 | 415 | 91 | ||
More Information
Set Membership: |
|

mcl1a
Genomic Map