Assay Details
| Gene Symbol: slc34a2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131624.1 | NP_571699.1 | 10 - 11 | 1344 | 65 | |
| GenBank mRNA | AF121796.1 | - | 10 - 11 | 1344 | 65 | |
| BC096858.1 | - | 10 - 11 | 1348 | 65 | ||
More Information
Set Membership: |
|

slc34a2a
Genomic Map