Assay Details
| Gene Symbol: hand2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131626.3 | NP_571701.3 | 1 - 2 | 1011 | 65 | |
| GenBank mRNA | AF228334.1 | - | 1 - 2 | 781 | 65 | |
| BC083365.1 | - | 1 - 2 | 1020 | 65 | ||
| BC165015.1 | - | 1 - 2 | 564 | 65 | ||
More Information
Set Membership: |
|

hand2
Genomic Map