Assay Details
| Gene Symbol: cyp19a1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131642.2 | NP_571717.2 | 2 - 3 | 233 | 101 | |
| XM_021470523.1 | XP_021326198.1 | 2 - 3 | 587 | 101 | ||
| GenBank mRNA | AF183908.1 | - | 2 - 3 | 212 | 101 | |
| AF226619.1 | - | 2 - 3 | 216 | 101 | ||
| BC076104.1 | - | 2 - 3 | 230 | 101 | ||
More Information
Set Membership: |
|

cyp19a1b
Genomic Map