Assay Details
| Gene Symbol: cyp19a1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131642.2 | NP_571717.2 | 4 - 5 | 542 | 81 | |
| XM_021470523.1 | XP_021326198.1 | 4 - 5 | 902 | 81 | ||
| GenBank mRNA | AF226619.1 | - | 4 - 5 | 525 | 81 | |
| BC076104.1 | - | 4 - 5 | 539 | 81 | ||
More Information
Set Membership: |
|

cyp19a1b
Genomic Map