Assay Details
| Gene Symbol: cyp19a1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131642.2 | NP_571717.2 | 7 - 8 | 942 | 67 | |
| XM_021470523.1 | XP_021326198.1 | 7 - 8 | 1302 | 67 | ||
| GenBank mRNA | AF183908.1 | - | 7 - 8 | 921 | 67 | |
| AF226619.1 | - | 7 - 8 | 925 | 67 | ||
More Information
Set Membership: |
|

cyp19a1b
Genomic Map