Assay Details
| Gene Symbol: cyp19a1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131642.2 | NP_571717.2 | 8 - 9 | 1107 | 90 | |
| XM_021470523.1 | XP_021326198.1 | 8 - 9 | 1467 | 90 | ||
| GenBank mRNA | AF120031.2 | - | 1 - 2 | 90 | ||
| AF183908.1 | - | 8 - 9 | 90 | |||
| AF226619.1 | - | 8 - 9 | 1090 | 90 | ||
| BC076104.1 | - | 9 - 10 | 90 | |||
More Information
Set Membership: |
|

cyp19a1b
Genomic Map