Assay Details
| Gene Symbol: atp1a2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131683.1 | NP_571758.1 | 18 - 19 | 2532 | 61 | |
| GenBank mRNA | AF286373.1 | - | 18 - 19 | 2532 | 61 | |
| AY028628.1 | - | 18 - 19 | 2466 | 61 | ||
| BC163629.1 | - | 18 - 19 | 2454 | 61 | ||
More Information
Set Membership: |
|

atp1a2a
Genomic Map