Assay Details
| Gene Symbol: atp1a3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131684.2 | NP_571759.2 | 20 - 21 | 3301 | 127 | |
| GenBank mRNA | AF308599.1 | - | 20 - 21 | 3227 | 127 | |
| BC064703.1 | - | 20 - 21 | 3301 | 127 | ||
More Information
Set Membership: |
|

atp1a3a
Genomic Map