Assay Details
| Gene Symbol: atp1a3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131684.2 | NP_571759.2 | 2 - 3 | 386 | 73 | |
| GenBank mRNA | AF308599.1 | - | 2 - 3 | 312 | 73 | |
| AW281430.1 | - | 2 - 3 | 452 | 73 | ||
| BC064703.1 | - | 2 - 3 | 386 | 73 | ||
| BC065650.1 | - | 2 - 3 | 439 | 73 | ||
More Information
Set Membership: |
|

atp1a3a
Genomic Map