Assay Details
| Gene Symbol: atp1a1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131690.1 | NP_571765.1 | 10 - 11 | 1483 | 65 | |
| GenBank mRNA | AY008375.1 | - | 10 - 11 | 1483 | 65 | |
| BC085663.1 | - | 10 - 11 | 1548 | 65 | ||
More Information
Set Membership: |
|

atp1a1b
Genomic Map