Assay Details
| Gene Symbol: tle3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131780.1 | NP_571855.1 | 15 - 16 | 1635 | 65 | |
| GenBank mRNA | AB040707.1 | - | 15 - 16 | 1635 | 65 | |
| BC162571.1 | - | 15 - 16 | 1627 | 65 | ||
| U96451.1 | - | 7 - 8 | 876 | 65 | ||
| Y12467.1 | - | 876 | 65 | |||
More Information
Set Membership: |
|

tle3b
Genomic Map