Assay Details
| Gene Symbol: tle3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131780.1 | NP_571855.1 | 8 - 9 | 674 | 66 | |
| XM_005168834.4 | XP_005168891.1 | 9 - 10 | 1161 | 66 | ||
| XM_009303189.3 | XP_009301464.1 | 9 - 10 | 1649 | 66 | ||
| XM_009303190.3 | XP_009301465.1 | 10 - 11 | 1282 | 66 | ||
| XM_009303191.3 | XP_009301466.1 | 10 - 11 | 1197 | 66 | ||
| XM_009303193.3 | XP_009301468.1 | 9 - 10 | 1646 | 66 | ||
| XM_009303194.3 | XP_009301469.1 | 9 - 10 | 1646 | 66 | ||
| XM_009303195.3 | XP_009301470.1 | 9 - 10 | 1650 | 66 | ||
| XM_009303196.3 | XP_009301471.1 | 9 - 10 | 1643 | 66 | ||
| XM_009303198.3 | XP_009301473.1 | 9 - 10 | 1616 | 66 | ||
| XM_009303199.3 | XP_009301474.1 | 8 - 9 | 1613 | 66 | ||
| XM_009303200.3 | XP_009301475.1 | 8 - 9 | 1617 | 66 | ||
| XM_009303201.3 | XP_009301476.1 | 8 - 9 | 1586 | 66 | ||
| XM_009303202.3 | XP_009301477.1 | 8 - 9 | 1583 | 66 | ||
| XM_009303203.3 | XP_009301478.1 | 8 - 9 | 1587 | 66 | ||
| XM_009303204.3 | XP_009301479.1 | 8 - 9 | 1584 | 66 | ||
| GenBank mRNA | AB040707.1 | - | 8 - 9 | 674 | 66 | |
| BC162571.1 | - | 8 - 9 | 666 | 66 | ||
More Information
Set Membership: |
|

tle3b
Genomic Map