Assay Details
| Gene Symbol: bcl2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131807.1 | NP_571882.1 | 2 - 3 | 801 | 74 | |
| XM_005162026.4 | XP_005162083.1 | 2 - 3 | 979 | 74 | ||
| GenBank mRNA | AF317837.1 | - | 2 - 3 | 801 | 74 | |
| BC044130.1 | - | 2 - 3 | 807 | 74 | ||
| BC071291.1 | - | 2 - 3 | 833 | 74 | ||
| BC165156.1 | - | 1 - 2 | 607 | 74 | ||
| DQ022560.1 | - | 2 - 3 | 715 | 74 | ||
More Information
Set Membership: |
|

bcl2l1
Genomic Map